missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRS1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17509-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
XPOT is a protein belonging to the RAN-GTPase exportin family that mediates export of tRNA from the nucleus to the cytoplasm. Translocation of tRNA to the cytoplasm occurs once exportin has bound both tRNA and GTP-bound RAN.Specifications
IRS1 | |
Polyclonal | |
Unconjugated | |
IRS1 | |
HIRS 1, insulin receptor substrate 1, IRS 1, IRS1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3667 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P35568 | |
IRS1 | |
IRS1 Fusion Protein Ag11714 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |