missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals ISIS Core Kit, 10 mm dNTPs
Ideal for high-fidelity amplification.
Brand: MP Biomedicals 11EPSISK100
Additional Details : Weight : 0.00001kg
Description
- Use to amplify DNA with difficult to read secondary structures.
- Optimal for blunt-end PCR cloning.
- The most thermostable polymerase available.
- Useful to amplify GC-rich sequences.
- Key applications: Molecular biology, PCR/amplification, PCR, cDNA synthesis, RT-PCR, DNA labeling, hIgh-fidelity PCR.
- Research Use Only (RUO).
Specifications
Store at −20°C | |
Standard PCR, cDNA Synthesis, RT-PCR, Nucleic Acid Labeling, High-fidelity PCR | |
100 U Isis™ DNA Pol at 1 U/µL, dNTPs at 10 mM each, Isis™ buffer 10xC (1 mL) with MgSO4, 1 mL MgSO4 at 30 mM, dNTPs 10 mM |
100 U | |
1 U/μL |