missing translation for 'onlineSavingsMsg'
Learn More

ISL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 18659-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16888554

  • 4721.67 SEK / 150µL
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ISL2 is a transcriptional factor that defines subclasses of motoneurons that segregate into columns in the spinal cord and select distinct axon pathways.
Specifications

Specifications

ISL2
Polyclonal
Unconjugated
ISL2
ISL LIM homeobox 2, ISL2, Islet 2
Rabbit
Antigen Affinity Chromatography
RUO
104360, 57233, 64843
-20°C
Liquid
Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P50480, Q96A47, Q9CXV0
Isl2
ISL2 Fusion Protein Ag13333
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers