missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ISM2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21540-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene contains a type 1 thrombospondin domain, which is present in thrombospondin, a number of proteins involved in the complement pathway, as well as in extracellular matrix proteins. Two alternatively spliced transcript variants encoding distinct isoforms have been observed.Specifications
ISM2 | |
Polyclonal | |
Unconjugated | |
ISM2 | |
ISM2, Isthmin 2, isthmin 2 homolog (zebrafish), TAIL1, THSD3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
145501, 217738 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6H9L7 | |
Ism2 | |
ISM2 Fusion Protein Ag16187 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |