Learn More
Iso-Amyl Acetate, 99+%, Certified AR for Analysis, Mix Of 2- & 3-Methylbutyl Acetate, Fisher Chemical™
Catalog Number p-7072841
Click to view available options
Quantity:
500 mL
2.5 L
123-92-2
C7H14O2
8946
123-92-2
C7H14O2
8946
Melting Point -78°C
Color Colorless
Boiling Point 142°C
Packaging Amber glass bottle
Quantity 500 mL
Vapor Density 4.49
Formula Weight 130.19g/mol
Vapor Pressure 4mmHg at 20°C
Physical Form Liquid
compliance-icons
Product Identifier
  • Isoamyl acetate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.