Läs mer
Iso E Super (Mixture of Isomers) (>80%), TRC
Produktkod. 30005653
Change view
Kvantitet:
10 mg
100 mg
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005653 100 mg
30004139 50 g
17951965 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30005653 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI815500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 54464-57-2
Molekylformel C16 H26 O
Molekylvikt (g/mol) 234.38
Synonym 1-(1,2,3,4,5,6,7,8-Octahydro-2,3,8,8-tetramethyl-2-naphthalenyl)ethanone, 1,2,3,4,5,6,7,8-Octahydro-2,3,8,8-tetramethyl-2-acetonaphthone, 1-(1,2,3,4,5,6,7,8-Octahydro-2,3,8,8-tetramethyl-2-naphthalenyl)ethan-1-one, 1-(1,2,3,4,5,6,7,8-Octahydro-2,3,8,8-tetramethyl-2-naphthyl)ethan-1-one, 1-(2,3,8,8-Tetramethyl-1,2,3,4,5,6,7,8-octahydronaphthalen-2-yl)ethanone, 1-(2,3,8,8-Tetramethyl-1,3,4,5,6,7-hexahydronaphthalen-2-yl)ethanone, 1',2',3',4',5',6',7',8'-Octahydro-2',3',8',8'-tetramethyl-2'-acetonaphthone, 2-Acetyl-2,3,8,8-tetramethyloctalin, 7-Acetyl-1,2,3,4,5,6,7,8-octahydro-1,1,6,7-tetramethylnaphthalene, Amberonne, Ambralux, Ambrotone, Iso Ambois Super, Iso-E Super, Isocyclemone, Isocyclemone E, OTNE, Orbitone, Ethanone, 1-(1,2,3,4,5,6,7,8-octahydro-2,3,8,8-tetramethyl-2-naphthalenyl)-, Iso E Super
IUPAC-namn 1-(2,3,8,8-tetramethyl-1,3,4,5,6,7-hexahydronaphthalen-2-yl)ethanone
LEDER CC1CC2=C(CC1(C)C(=O)C)C(C)(C)CCC2
Specifikationer
Kemiskt namn eller material Iso E Super
Kvantitet 100 mg
InChI formel InChI=1S/C16H26O/c1-11-9-13-7-6-8-15(3,4)14(13)10-16(11,5)12(2)17/h11H,6-10H2,1-5H3
Formel vikt 234.1984