Läs mer
(+/-)-Iso Myosmine, Technical Grade, TRC
Produktkod. 30180275
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180275 5 mg
30179967 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30180275 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI8213505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 53844-46-5
Molekylformel C9H10N2
Molekylvikt (g/mol) 146.19
Synonym 3-(3,4-Dihydro-2H-pyrrol-2-yl)pyridine, (+/-))-Isomyosamine;
LEDER C1(C2=CC=CN=C2)CCC=N1
Specifikationer
Kemiskt namn eller material (+/-)-Iso Myosmine
Analyt- eller komponentnamn (+/-)-Iso Myosmine
Frakt skick Dry ice
Industry Type Forensics