Learn More
Isobornyl methacrylate, 85-90%, stabilized, Thermo Scientific Chemicals
Catalog Number p-150370
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
7534-94-3
C14H22O2
222.33
MFCD00081070
IAXXETNIOYFMLW-UHFFFAOYNA-N
ibma, isobornyl methacrylate, methacrylic acid isobornyl ester, iso-bornyl methacrylate, isobornyl methacrylate, technical grade, isobornyl methacrylate, stabilized with mehq, exo-1,7,7-trimethylbicyclo 2.2.1 hept-, 2-yl methacrylate, 1r,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-yl 2-methylprop-2-enoate
71311141
[(1R,4R)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] 2-methylprop-2-enoate
CC(=C)C(=O)OC1CC2CCC1(C)C2(C)C
7534-94-3
C14H22O2
222.33
MFCD00081070
IAXXETNIOYFMLW-UHFFFAOYNA-N
ibma, isobornyl methacrylate, methacrylic acid isobornyl ester, iso-bornyl methacrylate, isobornyl methacrylate, technical grade, isobornyl methacrylate, stabilized with mehq, exo-1,7,7-trimethylbicyclo 2.2.1 hept-, 2-yl methacrylate, 1r,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-yl 2-methylprop-2-enoate
71311141
[(1R,4R)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] 2-methylprop-2-enoate
CC(=C)C(=O)OC1CC2CCC1(C)C2(C)C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point -60.0°C
Color Colorless to Yellow
Density 0.9800g/mL
Boiling Point 245.0°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 85% min. (GC)
Packaging Glass bottle
Refractive Index 1.4760 to 1.4780
Quantity 50 g
Specific Gravity 0.98
Solubility Information Solubility in water: insoluble
Stabilizer 100 to 200ppm MEHQ
Formula Weight 222.33
Percent Purity 85-90%
Physical Form Liquid
Chemical Name or Material Isobornyl methacrylate, 85-90%
Show More Show Less
compliance-icons
Product Identifier
  • Isobornyl methacrylate, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.