missing translation for 'onlineSavingsMsg'
Learn More

Isobutanesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog Number 15461559
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
35432-36-1
C4H9ClO2S
156.624
MFCD07369800
NQLNHTOCFWUYQE-UHFFFAOYSA-N
isobutanesulfonyl chloride, isobutylsulfonyl chloride, 1-propanesulfonyl chloride, 2-methyl, 2-methylpropane-1-sulfonylchloride, 2-methyl-1-propylsulfonyl chloride, 2-methyl-1-propanesulfonyl chloride, 2-methyl-propane-1-sulfonyl chloride, isobutane sulfonyl chloride, isobutane-1-sulfonylchloride, acmc-1ae06
182236
2-methylpropane-1-sulfonyl chloride
CC(C)CS(=O)(=O)Cl
This item is not returnable. View return policy
35432-36-1
C4H9ClO2S
156.624
MFCD07369800
NQLNHTOCFWUYQE-UHFFFAOYSA-N
isobutanesulfonyl chloride, isobutylsulfonyl chloride, 1-propanesulfonyl chloride, 2-methyl, 2-methylpropane-1-sulfonylchloride, 2-methyl-1-propylsulfonyl chloride, 2-methyl-1-propanesulfonyl chloride, 2-methyl-propane-1-sulfonyl chloride, isobutane sulfonyl chloride, isobutane-1-sulfonylchloride, acmc-1ae06
182236
2-methylpropane-1-sulfonyl chloride
CC(C)CS(=O)(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.20 g/mL
Flash Point 88°C (190°F)
Linear Formula (CH3)2CHCH2SO2Cl
Refractive Index 1.454
Quantity 250 mg
UN Number UN3265
Beilstein 1748728
Sensitivity Moisture sensitive
Formula Weight 156.62
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Isobutanesulfonyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'