Läs mer
Isobutyl Acetoacetate, TRC
Produktkod. 30403448
Change view
Kvantitet:
50 g
Produktkod. Kvantitet
30403448 50 g
1 options
Produktkod. 30403448 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI78004550G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Isobutyl Acetoacetate
CAS 7779-75-1
Molekylformel C8H14O3
Kvantitet 50 g
Synonym 2-Methylpropyl-3-oxobutanoate, Isobutyl-3-oxobutanoate, Isobutyl Ester Acetoacetic Acid
InChI formel InChI=1S/C8H14O3/c1-6(2)5-11-8(10)4-7(3)9/h6H,4-5H2,1-3H3
LEDER CC(C)COC(=O)CC(=O)C
Molekylvikt (g/mol) 158.19
Formel vikt 158.0943