Läs mer
Isobutyl Ethyl Ether, TRC
Produktkod. 30404971
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30404971 500 mg
1 options
Produktkod. 30404971 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI627050500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Ethoxy-2-methylpropane
CAS 627-02-1
Molekylformel C6 H14 O
Kvantitet 500 mg
Synonym Isobutyl Ethyl Ether, Propane, 1-ethoxy-2-methyl-, Ether, ethyl isobutyl (6CI,7CI,8CI), 1-Ethoxy-2-methylpropane, Ethyl iso-butyl ether, Ethyl isobutyl ether, Isobutyl ethyl ether
InChI formel InChI=1S/C6H14O/c1-4-7-5-6(2)3/h6H,4-5H2,1-3H3
LEDER CCOCC(C)C
IUPAC-namn 1-ethoxy-2-methylpropane
Molekylvikt (g/mol) 102.175
Formel vikt 102.104