Läs mer
Isocaffeine, TRC
Produktkod. 17932645
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17932645 100 mg
17912295 1 g
17924785 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17932645 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI789200100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 519-32-4
Molekylformel C8 H10 N4 O2
Molekylvikt (g/mol) 194.19
Synonym 1,3,9-Trimethyl-3,9-dihydro-1H-purine-2,6-dione, Isocaffeine, Caffeine Imp. C (EP)
IUPAC-namn 1,3,9-trimethylpurine-2,6-dione
LEDER CN1C(=O)N(C)c2c(ncn2C)C1=O
Specifikationer
Kemiskt namn eller material 1,3,9-Trimethyl-3,9-dihydro-1H-purine-2,6-dione (Isocaffeine)
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C8H10N4O2/c1-10-4-9-5-6(10)11(2)8(14)12(3)7(5)13/h4H,1-3H3
Formel vikt 194.0804
Procent renhet >95%