missing translation for 'onlineSavingsMsg'
Läs mer

Isocarboxazid (1.0 mg/mL in Acetonitrile), TRC

Produktkod. 30464792
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30464792 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30464792 Leverantör Toronto Research Chemicals Leverantörsnummer TRCKIT83455X1ML

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Isocarboxazid
CAS 59-63-2
Molekylformel C12 H13 N3 O2
Kvantitet 5 x 1 mL
Synonym 3-Isoxazolecarboxylic acid, 5-methyl-, 2-(phenylmethyl)hydrazide3-Isoxazolecarboxylic acid, 5-methyl-, 2-benzylhydrazide (6CI,7CI,8CI), 1-Benzyl-2-(5-methyl-3-isoxazolyl-carbonyl)hydrazine, 5-Methyl-3-isoxazolecarboxylic acid 2-benzylhydrazide, BMIH, Benazide, Enerzer, Isocarboxazid, Isocarboxazide, Isocarboxyzid, Marplan, Marplon, NSC 169893, N'-Benzyl N-methyl-5-isoxazolecarboxylhydrazide-3, Ro 5-0831
InChI formel InChI=1S/C12H13N3O2/c1-9-7-11(15-17-9)12(16)14-13-8-10-5-3-2-4-6-10/h2-7,13H,8H2,1H3,(H,14,16)
LEDER Cc1onc(c1)C(=O)NNCc2ccccc2
IUPAC-namn N'-benzyl-5-methyl-1,2-oxazole-3-carbohydrazide
Molekylvikt (g/mol) 231.25
Formel vikt 231.1008
Koncentration eller sammansättning (efter analyt eller komponenter) Isocarboxazid 1 mg/mL
Visa mer Visa mindre
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.