Läs mer
Isoconazole Nitrate, TRC
Produktkod. 30407461
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30407461 1 g
1 options
Produktkod. 30407461 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI7982501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Isoconazole Nitrate
CAS 24168-96-5
Anteckningar om renhetsgrad HPLC
Molekylformel C18 H14 Cl4 N2 O . H N O3
Kvantitet 1 g
Synonym Isoconazole Nitrate, 1-[(2RS)-2-[(2,6-Dichlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate, Miconazole Nitrate Imp. D (EP) as Nitrate
InChI formel InChI=1S/C18H14Cl4N2O.HNO3/c19-12-4-5-13(17(22)8-12)18(9-24-7-6-23-11-24)25-10-14-15(20)2-1-3-16(14)21;2-1(3)4/h1-8,11,18H,9-10H2;(H,2,3,4)
LEDER O[N+](=O)[O-].Clc1ccc(C(Cn2ccnc2)OCc3c(Cl)cccc3Cl)c(Cl)c1
IUPAC-namn 1-[2-(2,4-dichlorophenyl)-2-[(2,6-dichlorophenyl)methoxy]ethyl]imidazole;nitric acid
Molekylvikt (g/mol) 479.14
Formel vikt 476.9817