missing translation for 'onlineSavingsMsg'
Läs mer

Isoconazole, TRC

Produktkod. 30393538
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30393538 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30393538 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI2127501MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Isoconazole
CAS 27523-40-6
Anteckningar om renhetsgrad HPLC
Molekylformel C18 H14 Cl4 N2 O
Kvantitet 1 mg
Synonym Imidazole, 1-[2,4-dichloro-β-[(2,6-dichlorobenzyl)oxy]phenethyl]- (8CI), 1-[2-(2,4-Dichlorophenyl)-2-[(2,6-dichlorophenyl)methoxy]ethyl]-1H-imidazole, (±)-Isconazole, Isoconazole
InChI formel InChI=1S/C18H14Cl4N2O/c19-12-4-5-13(17(22)8-12)18(9-24-7-6-23-11-24)25-10-14-15(20)2-1-3-16(14)21/h1-8,11,18H,9-10H2
LEDER Clc1ccc(C(Cn2ccnc2)OCc3c(Cl)cccc3Cl)c(Cl)c1
IUPAC-namn 1-[2-(2,4-dichlorophenyl)-2-[(2,6-dichlorophenyl)methoxy]ethyl]imidazole
Molekylvikt (g/mol) 416.13
Formel vikt 413.986
Procent renhet >95
Visa mer Visa mindre
Produkttitel
Välj ett problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.