missing translation for 'onlineSavingsMsg'
Läs mer

(-)-Isocorypalmine, TRC

Produktkod. 30011430
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30011430 2.5 mg 2.5mg
30005968 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30011430 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI7980002.5MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 483-34-1
Molekylformel C20 H23 N O4
Molekylvikt (g/mol) 341.4
Synonym (13aS)-5,8,13,13a-Tetrahydro-3,9,10-trimethoxy-6H-dibenzo[a,g]quinolizin-2-ol, Tetrahydro-columbamine, 2-O-Desmethyl-L-THP, Corydalis L, O10-Methylstepholidine, Tetrahydrocolumbamine
IUPAC-namn (13aS)-3,9,10-trimethoxy-6,8,13,13a-tetrahydro-5H-isoquinolino[3,2-a]isoquinolin-2-ol
LEDER COc1cc2CCN3Cc4c(C[C@H]3c2cc1O)ccc(OC)c4OC

Specifikationer

Kemiskt namn eller material (-)-Isocorypalmine
Kvantitet 2.5 mg
InChI formel InChI=1S/C20H23NO4/c1-23-18-5-4-12-8-16-14-10-17(22)19(24-2)9-13(14)6-7-21(16)11-15(12)20(18)25-3/h4-5,9-10,16,22H,6-8,11H2,1-3H3/t16-/m0/s1
Formel vikt 341.1627
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.