Läs mer
Isofagomine D-Tartrate, TRC
Produktkod. 30010557
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010557 50 mg
30011284 5 mg
30012120 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010557 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI81601050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 957230-65-8
Molekylformel C6 H13 N O3 . C4 H6 O6
Molekylvikt (g/mol) 297.26
IUPAC-namn (2S,3S)-2,3-dihydroxybutanedioic acid;(3R,4R,5R)-5-(hydroxymethyl)piperidine-3,4-diol
LEDER OC[C@H]1CNC[C@@H](O)[C@@H]1O.O[C@@H]([C@H](O)C(=O)O)C(=O)O
Specifikationer
Kemiskt namn eller material Isofagomine D-Tartrate
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C6H13NO3.C4H6O6/c8-3-4-1-7-2-5(9)6(4)10;5-1(3(7)8)2(6)4(9)10/h4-10H,1-3H2;1-2,5-6H,(H,7,8)(H,9,10)/t4-,5-,6-;1-,2-/m10/s1
Formel vikt 297.106
Procent renhet >95%