Läs mer
Isohumulone, TRC
Produktkod. 30004134
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004134 5 mg
30006776 25 mg
30005003 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30004134 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI7904205MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 25522-96-7
Molekylformel C21 H30 O5
Molekylvikt (g/mol) 362.46
Synonym 3,4-Dihydroxy-5-(3-methyl-2-buten-1-yl)-2-(3-methyl-1-oxobutyl)-4-(4-methyl-1-oxo-3-penten-1-yl)-2-cyclopenten-1-one, Iso-n-humulone, IsoHopCO2N
IUPAC-namn 3,4-dihydroxy-2-(3-methylbutanoyl)-5-(3-methylbut-2-enyl)-4-(4-methylpent-3-enoyl)cyclopent-2-en-1-one
LEDER CC(C)CC(=O)C1=C(O)C(O)(C(CC=C(C)C)C1=O)C(=O)CC=C(C)C
Specifikationer
Kemiskt namn eller material Isohumulone
Kvantitet 5 mg
InChI formel InChI=1S/C21H30O5/c1-12(2)7-9-15-19(24)18(16(22)11-14(5)6)20(25)21(15,26)17(23)10-8-13(3)4/h7-8,14-15,25-26H,9-11H2,1-6H3
Formel vikt 362.2093