Learn More
Isoindoline hydrochloride, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 32372-82-0 |
| Molecular Formula | C8H10ClN |
| MDL Number | MFCD03502377 |
| Quantity | 5 g |
| SMILES | C12=C(CNC2)C=CC=C1.[H]Cl |
| Molecular Weight (g/mol) | 155.62 |
| Percent Purity | 97% |
| Chemical Name or Material | Isoindoline hydrochloride |