Läs mer
Isoleukotoxin Diol, TRC
Produktkod. 30009398
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30009398 10 mg
1 options
Produktkod. 30009398 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI82023010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Isoleukotoxin Diol
CAS 73889-55-1
Molekylformel C18H34O4
Kvantitet 10 mg
Synonym (9Z,12R,13R)-rel-12,13-Dihydroxy-9-octadecenoic Acid, [R*,R*-(Z)]-12,13-Dihydroxy-9-octadecenoic Acid, rel-(9Z,12R,13R)-12,13-Dihydroxy-9-octadecenoic Acid;
InChI formel InChI=1S/C18H34O4/c1-2-3-10-13-16(19)17(20)14-11-8-6-4-5-7-9-12-15-18(21)22/h8,11,16-17,19-20H,2-7,9-10,12-15H2,1H3,(H,21,22)/b11-8-/t16-,17-/m0/s1
LEDER CCCCC[C@H](O)[C@@H](O)C\C=C/CCCCCCCC(=O)O
Molekylvikt (g/mol) 314.46
Formel vikt 314.2457