Läs mer
Isometamidium Bromide Hydrobromide, TRC
Produktkod. 17948994
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17948994 1 mg
17969554 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17948994 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI8212651MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4174-69-0
Molekylformel C28 H26 N7 . Br . Br H
Molekylvikt (g/mol) 621.37
Synonym Isometamidium Bromide Hydrobromide, Phenanthridinium, 3-amino-8-[3-[3-(aminoiminomethyl)phenyl]-2-triazen-1-yl]-5-ethyl-6-phenyl-, bromide, hydrobromide (1:1:1), Phenanthridinium, 8-[3-(m-amidinophenyl)-2-triazeno]-3-amino-5-ethyl-6-phenyl-, bromide, hydrobromide (8CI), M and B 4180
IUPAC-namn 3-[(2E)-2-(3-amino-5-ethyl-6-phenylphenanthridin-5-ium-8-yl)iminohydrazinyl]benzenecarboximidamide;bromide;hydrobromide
LEDER Br.[Br-].CC[n+]1c(c2ccccc2)c3cc(ccc3c4ccc(N)cc14)N=NNc5cccc(c5)C(=N)N
Specifikationer
Kemiskt namn eller material Isometamidium bromide hydrobromide
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C28H25N7.2BrH/c1-2-35-26-16-20(29)11-13-24(26)23-14-12-22(17-25(23)27(35)18-7-4-3-5-8-18)33-34-32-21-10-6-9-19(15-21)28(30)31;;/h3-17,29H,2H2,1H3,(H4,30,31,32,33);2*1H
Formel vikt 619.0695
Procent renhet >95%