Läs mer
Isometamidium Chloride Hydrochloride, TRC
Produktkod. 17972755
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17972755 1 mg
17915115 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17972755 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI8212751MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6798-24-9
Molekylformel C28 H26 N7 . Cl . Cl H
Molekylvikt (g/mol) 532.47
Synonym Phenanthridinium, 3-amino-8-[3-[3-(aminoiminomethyl)phenyl]-2-triazen-1-yl]-5-ethyl-6-phenyl-, chloride, hydrochloride (1:1:1), 8-[3-(m-Amidinophenyl)-2-triazeno]-3-amino-5-ethyl-6-phenylphenanthridinium chloride, hydrochloride (7CI), Isometamidium hydrochloride (6CI), Phenanthridinium, 3-amino-8-[3-[3-(aminoiminomethyl)phenyl]-1-triazenyl]-5-ethyl-6-phenyl-, chloride, monohydrochloride (9CI), Phenanthridinium, 8-[3-(m-amidinophenyl)-2-triazeno]-3-amino-5-ethyl-6-phenyl-, chloride, monohydrochloride (8CI), Isometamidium chloride hydrochloride, Samorin, Trypamidium, veridium
IUPAC-namn 3-[2-(3-amino-5-ethyl-6-phenylphenanthridin-5-ium-8-yl)iminohydrazinyl]benzenecarboximidamide;chloride;hydrochloride
LEDER Cl.[Cl-].CC[n+]1c(c2ccccc2)c3cc(ccc3c4ccc(N)cc14)N=NNc5cccc(c5)C(=N)N
Specifikationer
Kemiskt namn eller material Isometamidium Chloride Hydrochloride
Kvantitet 1 mg
InChI formel InChI=1S/C28H25N7.2ClH/c1-2-35-26-16-20(29)11-13-24(26)23-14-12-22(17-25(23)27(35)18-7-4-3-5-8-18)33-34-32-21-10-6-9-19(15-21)28(30)31;;/h3-17,29H,2H2,1H3,(H4,30,31,32,33);2*1H
Formel vikt 531.1705