Läs mer
Isononyl Acrylate (mixture of branched chain isomers, stabilized with MEHQ) (Technical Grade), TRC
Produktkod. 30408514
Change view
Kvantitet:
100 mL
Produktkod. Kvantitet
30408514 100 mL
1 options
Produktkod. 30408514 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI821818100ML
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Isononyl Acrylate (mixture of branched chain isomers) (Technical Grade)
CAS 51952-49-9
Molekylformel C12H22O2
Kvantitet 100 mL
InChI formel InChI=1S/C12H22O2/c1-4-12(13)14-10-8-6-5-7-9-11(2)3/h4,11H,1,5-10H2,2-3H3
LEDER CC(C)CCCCCCOC(=O)C=C
IUPAC-namn 7-methyloctyl prop-2-enoate
Molekylvikt (g/mol) 198.31
Formel vikt 198.162