Läs mer
Isopropyl 3-Mercaptopropionate, TRC
Produktkod. 30240961
Change view
Kvantitet:
250 mg
500 mg
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30240961 250 mg
30240778 2.5 g
30240482 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30240961 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI872908250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7383-64-4
Molekylformel C6H12O2S
Molekylvikt (g/mol) 148.22
IUPAC-namn propan-2-yl 3-sulfanylpropanoate
LEDER CC(C)OC(=O)CCS
Specifikationer
Kemiskt namn eller material Isopropyl 3-Mercaptopropionate
Kvantitet 250 mg
InChI formel InChI=1S/C6H12O2S/c1-5(2)8-6(7)3-4-9/h5,9H,3-4H2,1-2H3
Formel vikt 148.0558