missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl DL-Mandelate, TRC
Specifications
Specifications
| Kemiskt namn eller material | Isopropyl Mandelate |
| CAS | 4118-51-8 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C11 H14 O3 |
| Kvantitet | 25 mg |
| Synonym | α-Hydroxy-benzeneacetic acid 1-Methylethyl ester, Propan-2-yl (2RS)-2-Hydroxy-2-phenylacetate, Mandelic acid, isopropyl ester (7CI,8CI), (±)-Isopropyl mandelate, (±)-Mandelic acid isopropyl ester, Isopropyl 2-hydroxy-2-phenylacetate, Isopropyl mandelate, NSC 6582 |
| InChI formel | InChI=1S/C11H14O3/c1-8(2)14-11(13)10(12)9-6-4-3-5-7-9/h3-8,10,12H,1-2H3 |
| LEDER | CC(C)OC(=O)C(O)c1ccccc1 |
| IUPAC-namn | propan-2-yl 2-hydroxy-2-phenylacetate |
| Molekylvikt (g/mol) | 194.227 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?