missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Isopropyl Ester of Cetirizine, Mikromol™
Beskrivning
Isopropyl Ester of Cetirizine
Specifikationer
Specifikationer
| Kemiskt namn eller material | Isopropyl Ester of Cetirizine |
| CAS | 324047-27-0 |
| Analyt- eller komponentnamn | Isopropyl Ester of Cetirizine |
| Frakt skick | Ice pack (-20°C) |
| Molekylformel | C24 H31 Cl N2 O3 |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | Isopropyl Ester of Cetirizine |
| InChI formel | InChI=1S/C24H31ClN2O3/c1-19(2)30-23(28)18-29-17-16-26-12-14-27(15-13-26)24(20-6-4-3-5-7-20)21-8-10-22(25)11-9-21/h3-11,19,24H,12-18H2,1-2H3 |
| LEDER | CC(C)OC(=O)COCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3 |
| IUPAC-namn | propan-2-yl 2-[2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?