Läs mer
Isopropyl Hydrogen Methylphosphonate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Isopropyl methylphosphonic acid |
| CAS | 1832-54-8 |
| Molekylformel | C4 H11 O3 P |
| Kvantitet | 500 mg |
| Synonym | IMPA, Isopropyl Methylphosphonate, Isopropyl Methylphosphonic Acid, Monoisopropyl Methylphosphonate, O-Isopropyl Hydrogen Methylphosphonate, O-Isopropyl Methylphosphonic Acid, Mono(1-methylethyl) P-methyl-phosphonic Acid Ester |
| InChI formel | InChI=1S/C4H11O3P/c1-4(2)7-8(3,5)6/h4H,1-3H3,(H,5,6) |
| LEDER | CC(C)OP(=O)(C)O |
| IUPAC-namn | methyl(propan-2-yloxy)phosphinic acid |
| Molekylvikt (g/mol) | 138.1 |
| Formel vikt | 138.0446 |