Läs mer
Isopropyl p-Tosylate, TRC
Produktkod. 30136164
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136164 10 g
30008370 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136164 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI87380010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2307-69-9
Molekylformel C10 H14 O3 S
Molekylvikt (g/mol) 214.28
Synonym Benzenesulfonic acid, 4-methyl-, 1-methylethyl ester, p-Toluenesulfonic acid, isopropyl ester (6CI,7CI,8CI), 2-Propyl tosylate, 4-Methylbenzenesulfonic acid 1-methylethyl ester, Isopropyl 4-Methylbenzenesulfonate, Isopropyl p-methylbenzenesulfonate, Isopropyl p-toluenesulfonate, Isopropyl p-tosylate, Isopropyl tosylate, Toluene-4-sulfonic acid isopropyl ester
IUPAC-namn propan-2-yl 4-methylbenzenesulfonate
LEDER CC(C)OS(=O)(=O)c1ccc(C)cc1
Specifikationer
Kemiskt namn eller material Isopropyl p-Tosylate
Anteckningar om renhetsgrad HPLC
Kvantitet 10 g
InChI formel InChI=1S/C10H14O3S/c1-8(2)13-14(11,12)10-6-4-9(3)5-7-10/h4-8H,1-3H3
Formel vikt 214.0664
Procent renhet >95%