missing translation for 'onlineSavingsMsg'
Learn More

Isopropylbarbituric acid, 97%, Thermo Scientific™

Catalog Number 15206038
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
7391-69-7
C7H10N2O3
170.168
GFPIGNBQTXNNAG-UHFFFAOYSA-N
5-isopropylbarbituric acid, 2,4,6 1h,3h,5h-pyrimidinetrione, 5-1-methylethyl, 5-isopropylpyrimidine-2,4,6 1h,3h,5h-trione, 5-isopropyl barbituric acid, barbituric acid, 5-isopropyl, barbituric acid, 5-ipr, 2,6 1h,3h,5h-pyrimidinetrione, 5-1-methylethyl, 5-1-methylethyl pyrimidine-2,4,6 1h,3h,5h-trione
81874
5-propan-2-yl-1,3-diazinane-2,4,6-trione
CC(C)C1C(=O)NC(=O)NC1=O
This item is not returnable. View return policy
7391-69-7
C7H10N2O3
170.168
GFPIGNBQTXNNAG-UHFFFAOYSA-N
5-isopropylbarbituric acid, 2,4,6 1h,3h,5h-pyrimidinetrione, 5-1-methylethyl, 5-isopropylpyrimidine-2,4,6 1h,3h,5h-trione, 5-isopropyl barbituric acid, barbituric acid, 5-isopropyl, barbituric acid, 5-ipr, 2,6 1h,3h,5h-pyrimidinetrione, 5-1-methylethyl, 5-1-methylethyl pyrimidine-2,4,6 1h,3h,5h-trione
81874
5-propan-2-yl-1,3-diazinane-2,4,6-trione
CC(C)C1C(=O)NC(=O)NC1=O
Melting Point 215.0°C to 217.0°C
Formula Weight 170.17
Quantity 25 g
Chemical Name or Material Isopropylbarbituric acid
compliance-icons compliance-icons
Product Identifier
  • Isopropylbarbituric acid, 97%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'