Läs mer
Isopyridoxal Hydrochloride, TRC
Produktkod. 30134448
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134448 25 mg
30132763 50 mg
30010830 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134448 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI82410025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 90005-85-9
Molekylformel C8H9NO3 . HCl
Molekylvikt (g/mol) 203.62
Synonym 5-Hydroxy-4-(hydroxymethyl)-6-methyl-3-Pyridinecarboxaldehyde Hydrochloride
IUPAC-namn 5-hydroxy-4-(hydroxymethyl)-6-methylnicotinaldehyde hydrochloride
LEDER O=CC1=C(CO)C(O)=C(C)N=C1.Cl
Specifikationer
Kemiskt namn eller material Isopyridoxal Hydrochloride
Kvantitet 25 mg
InChI formel InChI=1S/C8H9NO3.ClH/c1-5-8(12)7(4-11)6(3-10)2-9-5;/h2-3,11-12H,4H2,1H3;1H
Formel vikt 203.04