Läs mer
Isorhamnetin-3-O-glucoside, MedChemExpress
Produktkod. 16431508
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
Förpackningsstorlek:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16431508 5 mg 5mg
16429098 10 mg -
16413906 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16431508 Leverantör Medchem Express Leverantörsnummer HYN0777
MedChemExpress Isorhamnetin-3-O-glucoside, a natural compound widely contained in many vegetables and rice, could be metabolized in intestinal microbiota after digestion.
Isorhamnetin-3-O-glucoside, a natural compound widely contained in many vegetables and rice, could be metabolized in intestinal microbiota after digestion.
Specifikationer
Kemiskt namn eller material Isorhamnetin-3-O-glucoside
CAS 5041-82-7
Färg Yellow
Anteckningar om renhetsgrad Research
Molekylformel C22H22O12
Kvantitet 5 mg
Löslighetsinformation DMSO : 250 mg/mL (522.58 mM; Need ultrasonic)
LEDER O=C1C(O[C@@H]([C@@H]([C@@H](O)[C@@H]2O)O)O[C@@H]2CO)=C(C3=CC(OC)=C(O)C=C3)OC4=CC(O)=CC(O)=C14
Molekylvikt (g/mol) 478.4
Formel vikt 478.4
Procent renhet 99.0%
Kvalitet Research
Fysisk form Powder
För användning med (applikation) Metabolism-protein/nucleotide metabolism