Learn More
Isosilybin, MedChemExpress
Product Code. 16441508
Encompass_Preferred
Change view
Click to view available options
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
16441508 5 mg
16439098 10 mg
16423906 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16441508 Supplier Medchem Express Supplier No. HYN0779
MedChemExpress Isosilybin (Isosilybinin) is a flavonoid from milk thistle; inhibits CYP3A4 induction with an IC50 of 74 μM.
Isosilybin (Isosilybinin) is a flavonoid from milk thistle
Specifications
Kemiskt namn eller material Isosilybin
CAS 72581-71-6
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C25H22O10
Kvantitet 5 mg
Synonym Isosilybinin
Löslighetsinformation DMSO : 100 mg/mL (207.28 mM; Need ultrasonic)
LEDER O=C1[C@H](O)[C@@H](C2=CC=C(OC(C3=CC=C(O)C(OC)=C3)C(CO)O4)C4=C2)OC5=CC(O)=CC(O)=C15
Molekylvikt (g/mol) 482.44
Formel vikt 482.44
Procent renhet 98.08%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease