missing translation for 'onlineSavingsMsg'
Learn More

Isovaleric acid, 99%, Thermo Scientific Chemicals

Catalog Number p-152260
Click to view available options
Quantity:
10 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
503-74-2
C5H10O2
102.13
MFCD00002726
GWYFCOCPABKNJV-UHFFFAOYSA-N
isovaleric acid, isopentanoic acid, 3-methylbutyric acid, delphinic acid, isopropylacetic acid, butanoic acid, 3-methyl, isovalerianic acid, isobutylformic acid, 3-methylbutyrate, beta-methylbutyric acid
10430
CHEBI:28484
3-methylbutanoic acid
CC(C)CC(O)=O
This item is not returnable. View return policy
503-74-2
C5H10O2
102.13
MFCD00002726
GWYFCOCPABKNJV-UHFFFAOYSA-N
isovaleric acid, isopentanoic acid, 3-methylbutyric acid, delphinic acid, isopropylacetic acid, butanoic acid, 3-methyl, isovalerianic acid, isobutylformic acid, 3-methylbutyrate, beta-methylbutyric acid
10430
CHEBI:28484
3-methylbutanoic acid
CC(C)CC(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -35.0°C
Color Colorless to Yellow
Density 0.9260g/mL
Boiling Point 176.0°C
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHCH2CO2H
Refractive Index 1.3990 to 1.4070
Quantity 10 g
Beilstein 02, 309
Merck Index 15, 5277
Specific Gravity 0.926
Solubility Information Solubility in water: 25g/L (20°C). Other solubilities: soluble in alcohol,chloroform and ether
Viscosity 2.4 mPa.s (20°C)
Formula Weight 102.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Isovaleric acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Isovaleric acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'