Läs mer
Isovaleric Acid Methyl Ester, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Methyl Isovalerate |
| CAS | 556-24-1 |
| Molekylformel | C6 H12 O2 |
| Kvantitet | 50 g |
| Synonym | Isovaleric acid, methyl ester (6CI,7CI,8CI), 3-Methylbutanoic acid methyl ester, Isopentanoic acid methyl ester, Methyl 3-methylbutanoate, Methyl 3-, Isovaleric Acid-Methyl Estermethylbutyrate, Methyl iso-valerate, Methyl isopentanoate, Methyl isovalerate |
| InChI formel | InChI=1S/C6H12O2/c1-5(2)4-6(7)8-3/h5H,4H2,1-3H3 |
| LEDER | COC(=O)CC(C)C |
| IUPAC-namn | methyl 3-methylbutanoate |
| Molekylvikt (g/mol) | 116.16 |
| Formel vikt | 116.0837 |