Läs mer
Isovitexin, TRC
Produktkod. 30132439
Change view
Kvantitet:
1 mg
5 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132439 50 mg
30137761 5 mg
17980095 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132439 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI91808550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 38953-85-4
Molekylformel C21 H20 O10
Molekylvikt (g/mol) 432.38
Synonym 4H-1-Benzopyran-4-one, 6-β-D-glucopyranosyl-5,7-dihydroxy-2-(4-hydroxyphenyl)-, Flavone, 6-β-D-glucopyranosyl-4',5,7-trihydroxy- (7CI), Homovitexin (6CI), 6-β-D-Glucopyranosyl-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 6-C-Glucosylapigenin, 6-C-β-D-Glucopyranosylapigenin, Apigenin 6-C-β-D-glucoside, Apigenin 6-C-β-glucopyranoside, Apigenin-6-C-glucoside, Apigenin-6-C-β-D-glucopyranoside, Avroside, Isoavroside, Isovitexin, Isovitexin (C-6 isomer), Saponaretin
IUPAC-namn 5,7-dihydroxy-2-(4-hydroxyphenyl)-6-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]chromen-4-one
LEDER OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)c2c(O)cc3OC(=CC(=O)c3c2O)c4ccc(O)cc4
Specifications
Kemiskt namn eller material Isovitexin
Kvantitet 50 mg
InChI formel InChI=1S/C21H20O10/c22-7-14-17(26)19(28)20(29)21(31-14)16-11(25)6-13-15(18(16)27)10(24)5-12(30-13)8-1-3-9(23)4-2-8/h1-6,14,17,19-23,25-29H,7H2/t14-,17-,19+,20-,21+/m1/s1
Formel vikt 432.1056