Läs mer
Isoxadifen, TRC
Produktkod. 17991995
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17991995 10 mg
17925055 50 mg
17941555 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17991995 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI91824810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 209866-92-2
Molekylformel C16 H13 N O3
Molekylvikt (g/mol) 267.28
Synonym Isoxadifen (free acid), 3-Isoxazolecarboxylic acid, 4,5-dihydro-5,5-diphenyl- (9CI, ACI), 4,5-Dihydro-5,5-diphenyl-3-isoxazolecarboxylic acid (ACI), Isoxadifen
IUPAC-namn 5,5-diphenyl-4H-1,2-oxazole-3-carboxylic acid
LEDER OC(=O)C1=NOC(C1)(c2ccccc2)c3ccccc3
Specifikationer
Kemiskt namn eller material Isoxadifen (free acid)
Kvantitet 10 mg
InChI formel InChI=1S/C16H13NO3/c18-15(19)14-11-16(20-17-14,12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10H,11H2,(H,18,19)
Formel vikt 267.0895