Läs mer
Isoxaflutole, TRC
Produktkod. 30406551
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30406551 10 mg
1 options
Produktkod. 30406551 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI91814010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Isoxaflutole
CAS 141112-29-0
Molekylformel C15 H12 F3 N O4 S
Kvantitet 10 mg
Synonym (5-Cyclopropyl-4-isoxazolyl)[2-(methylsulfonyl)-4-(trifluoromethyl)phenyl]methanone, Balance, Balance (herbicide), Balance Pro, EXP 30953, EXP 31130A, Isoxaflutole, Merlin, Provence, RP 201772, RPA 201772, Methanone, (5-cyclopropyl-4-isoxazolyl)[2-(methylsulfonyl)-4-(trifluoromethyl)phenyl]-
InChI formel InChI=1S/C15H12F3NO4S/c1-24(21,22)12-6-9(15(16,17)18)4-5-10(12)13(20)11-7-19-23-14(11)8-2-3-8/h4-8H,2-3H2,1H3
LEDER CS(=O)(=O)c1cc(ccc1C(=O)c2cnoc2C3CC3)C(F)(F)F
IUPAC-namn (5-cyclopropyl-1,2-oxazol-4-yl)-[2-methylsulfonyl-4-(trifluoromethyl)phenyl]methanone
Molekylvikt (g/mol) 359.32
Formel vikt 359.0439