Läs mer
Ispinesib, MedChemExpress
Produktkod. 16484017
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16484017 50 mg
16408108 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16484017 Leverantör Medchem Express Leverantörsnummer HY50759
MedChemExpress Ispinesib is a specific inhibitor of kinesin spindle protein (KSP), with a Ki app of 1.7 nM.
Ispinesib is a specific inhibitor of kinesin spindle protein (KSP), with a Ki app of 1.7 nM.
Specifikationer
Kemiskt namn eller material Ispinesib
Fysisk form Solid
CAS 336113-53-2
Färg Light Yellow
Kvantitet 50 mg
Anteckningar om renhetsgrad Research
Molekylformel C30H33ClN4O2
För användning med (applikation) Cancer-programmed cell death
Synonym SB-715992
Löslighetsinformation DMSO : ≥ 125 mg/mL (241.75 mM)
LEDER O=C1N(CC2=CC=CC=C2)C([C@@H](C(C)C)N(CCCN)C(C3=CC=C(C)C=C3)=O)=NC4=CC(Cl)=CC=C14
Molekylvikt (g/mol) 517.06
Formel vikt 517.06
Procent renhet 98.28%