Läs mer
Istaroxime hydrochloride, MedChemExpress
Produktkod. 16432847
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16432847 50 mg
16426768 10 mg
16429317 5 mg
16426896 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16432847 Leverantör Medchem Express Leverantörsnummer HY15718A
MedChemExpress Istaroxime hydrochloride is a Na+/K+-ATPase inhibitor (IC50=0.11 μM) and a sarcoplasmic/endoplasmic reticulum calcium ATPase 2 (SERCA 2) activator.
Istaroxime hydrochloride is a Na+/K+-ATPase inhibitor (IC50=0.11 μM) and a sarcoplasmic/endoplasmic reticulum calcium ATPase 2 (SERCA 2) activator.
Specifikationer
Kemiskt namn eller material Istaroxime hydrochloride
Fysisk form Solid
CAS 374559-48-5
Färg White
Kvantitet 50 mg
Anteckningar om renhetsgrad Research
Molekylformel C21H33ClN2O3
För användning med (applikation) COVID-19-immunoregulation
Synonym PST2744 hydrochloride
Löslighetsinformation DMSO : ≥ 45 mg/mL (113.36 mM) ∣H2O : 25 mg/mL (62.98 mM; Need ultrasonic)
LEDER O=C1CC[C@@]2([H])[C@]3([H])CC([C@@]4([H])C/C(CC[C@]4(C)[C@@]3([H])CC[C@@]21C)=N/OCCN)=O.[H]Cl
Molekylvikt (g/mol) 396.95
Formel vikt 396.95
Procent renhet 98.13%