missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITGA6X1A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1236.20 SEK - 3313.85 SEK
Specifications
Antigen | ITGA6X1A |
---|---|
Concentration | 0.17 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16839754
|
Proteintech
20231-1-AP-20UL |
20 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16829754
|
Proteintech
20231-1-AP-150UL |
150 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
E3 ubiquitin-protein ligase that mediates the ubiquitination of p53/TP53 in the late response to DNA damage, and acts as a positive regulator of p53/TP53 stability, thereby regulating the G1/S DNA damage checkpoint. May act by catalyzing the formation of short polyubiquitin chains on p53/TP53 that are not targeted to the proteasome. In response to ionizing radiation, interacts with MDM2 and enhances p53/TP53 ubiquitination, possibly by restricting MDM2 from extending polyubiquitin chains on ubiquitinated p53/TP53. Plays a role in RPA-mediated DNA damage signaling and repair.Specifications
ITGA6X1A | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P23229, Q61739 | |
16403, 3655 | |
Peptide | |
Primary | |
-20°C | |
ITGA6 |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CD49f, Integrin alpha 6, integrin, alpha 6, ITGA6, ITGA6B, VLA 6 | |
Itga6 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |