Läs mer
ITX5061, MedChemExpress
Produktkod. 16470418
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16470418 5 mg
16437958 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16470418 Leverantör Medchem Express Leverantörsnummer HY19900
MedChemExpress ITX5061 is a type II inhibitor of p38 MAPK and also an antagonist of scavenger receptor B1 (SR-B1).
ITX5061 is a type II inhibitor of p38 MAPK and also an antagonist of scavenger receptor B1 (SR-B1).
Specifikationer
Kemiskt namn eller material ITX5061
CAS 1252679-52-9
Färg Light Brown
Anteckningar om renhetsgrad Research
Molekylformel C30H38ClN3O7S
Kvantitet 5 mg
Löslighetsinformation DMSO : ≥ 83.3 mg/mL (134.32 mM)
LEDER O=C(NC1=CC(C(C)(C)C)=CC(NS(=O)(C)=O)=C1OC)C(C2=C3C=CC=CC3=C(OCCN4CCOCC4)C=C2)=O.Cl
Molekylvikt (g/mol) 620.16
Formel vikt 620.16
Procent renhet 98.38%
Kvalitet Research
Fysisk form Powder
För användning med (applikation) COVID-19-anti-virus