Läs mer
Ivermectin B1 Aglycon, TRC
Produktkod. 30004131
Change view
Kvantitet:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004131 25 mg
30009232 5 mg
17921735 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30004131 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI94080525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 73162-95-5
Molekylformel C34 H50 O8
Molekylvikt (g/mol) 586.76
Synonym Milbemycin B, 5-O-demethyl-28-deoxy-6,28-epoxy-13-hydroxy-25-[(1S)-1-methylpropyl]-, (6R,13S,25R)-, Ivermectin Imp. G (EP);(6R,13S,25R)-5-O-Demethyl-28-deoxy-6,28-epoxy-13-hydroxy-25-[(1S)-1-methylpropyl]-milbemycin B, [6R,13S,25R(S)]-5-O-Demethyl-28-deoxy-6,28-epoxy-13-hydroxy-25-(1-methylpropyl)-milbemycin B, Spiro[11,15-methano-2H,13H,17H-furo[4,3,2-pq][2,6]benzodioxacyclooctadecin-13,2'-[2H]pyran] Milbemycin B Deriv., 22,23-Dihydroavermectin B1a Aglycon, Ivermectin Aglycone
LEDER CC[C@H](C)[C@H]1O[C@]2(CC[C@@H]1C)C[C@@H]3C[C@@H](C\C=C(/C)\[C@@H](O)[C@@H](C)\C=C\C=C\4/CO[C@@H]5[C@H](O)C(=C[C@@H](C(=O)O3)[C@]45O)C)O2
Specifikationer
Kemiskt namn eller material Ivermectin B1 Aglycon
Kvantitet 25 mg
InChI formel InChI=1S/C34H50O8/c1-7-19(2)30-22(5)13-14-33(42-30)17-26-16-25(41-33)12-11-21(4)28(35)20(3)9-8-10-24-18-39-31-29(36)23(6)15-27(32(37)40-26)34(24,31)38/h8-11,15,19-20,22,25-31,35-36,38H,7,12-14,16-18H2,1-6H3/b9-8+,21-11+,24-10+/t19-,20-,22-,25+,26-,27-,28-,29+,30+,31+,33+,34+/m0/s1
Formel vikt 586.3506