missing translation for 'onlineSavingsMsg'
Learn More

IYD Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 16667-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16886364

  • 3665.00 SEK / 150µL
Estimated Shipment: 03-04-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes an enzyme that catalyzes the oxidative NADPH-dependent deiodination of mono- and diiodotyrosine, which are the halogenated byproducts of thyroid hormone production. The N-terminus of the protein functions as a membrane anchor. Mutations in this gene cause congenital hypothyroidism due to dyshormonogenesis type 4, which is also referred to as deiodinase deficiency, or iodotyrosine dehalogenase deficiency, or thyroid hormonogenesis type 4. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

IYD
Polyclonal
Unconjugated
IYD
C6orf71, DEHAL1, Iodotyrosine dehalogenase 1, iodotyrosine deiodinase, IYD, IYD 1
Rabbit
Antigen Affinity Chromatography
RUO
389434
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6PHW0
IYD
IYD Fusion Protein Ag10146
150 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers