Learn More
GPE Scientific™ J Young Spherical Joint, Female/O-Ring Cup Joint
GPE Scientific™ J Young Spherical Joint, Female/O-Ring Cup Joint is an interchangeable spherical joint available with Viton or PTFE-covered Viton O-Ring.
Brand:  GPE Scientific™ GBF/28/15
Additional Details : Weight : 0.09000kg
 View more versions of this product
Product Code. 15844754
  • 313.00 SEK / Each
Estimated Shipment: 17-07-2024
to see stock.
Add to basket
Description
Description
  • Spherical joint designed for use with ball joint
  • Available with different joint sizes, shank bores, and shank ODs
Specifications
Specifications
Shank Bore: 15 mm, Shank OD: 19 mm
Female Spherical Joint
28/15