Learn More
GPE Scientific™ J Young Spherical Joint, Female/O-Ring Cup Joint
GPE Scientific™ J Young Spherical Joint, Female/O-Ring Cup Joint is an interchangeable spherical joint available with Viton or PTFE-covered Viton O-Ring.
Brand:  GPE Scientific™ GBF/12/2
Additional Details : Weight : 0.08000kg
 View more versions of this product
Product Code. 15894744
  • 310.00 SEK / Each
Estimated Shipment: 01-08-2024
to see stock.
Add to basket
Description
Description
  • Spherical joint designed for use with ball joint
  • Available with different joint sizes, shank bores, and shank ODs
Specifications
Specifications
Shank Bore: 2 mm, Shank OD: 7 mm
Female Spherical Joint
12/2