missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1236.20 SEK - 3313.85 SEK
Specifications
Antigen | JAK2 |
---|---|
Concentration | 0.34 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16877604
|
Proteintech
17670-1-AP-20UL |
20 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16867604
|
Proteintech
17670-1-AP-150UL |
150 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Janus Activating Kinase 2 (JAK2) is a non-rececptor tyrosine kinase involved in various processes such as cell growth, development, differentiation, and histone modifications. In response to a varitety of cytokine or related factors (e.g., interferon, interleukins), JAKs are activated via phosphorylation at 2 adjacent tyrosine residues. The activation of JAKs can lead to the phosphorylation of STAT (signal transducers and activatiors of transcription) proteins, which dimerize and translocate to the nucleus. Once translocated to the nucleus, the STAT proteins can modify transcription of numerous genes, including interferon-stimulated genes. JAK2 is required for the IFN gamma-receptor complex initiation and JAK1 functions as an amplifier. Some studies have suggested that the role of JAK2 might be performed by Tyk2 and JAK3, if they were positioned correctly within the IFN gamma-receptor complex. Mutations affecting this gene can cause Budd-Chiari syndrome, Polycythemia vera, Thrombocythemia 3, myelofibrosis, or Acute myelogenous leukemia.Specifications
JAK2 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
O60674, Q62120, Q62689 | |
16452, 24514, 3717 | |
JAK2 Fusion Protein Ag11264 | |
Primary | |
-20°C | |
JAK2 |
0.34 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
JAK 2, JAK2, Janus kinase 2, JTK10, Tyrosine protein kinase JAK2 | |
Jak2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |