missing translation for 'onlineSavingsMsg'
Learn More

JAK2 Recombinant Human Protein, JH1 and JH2 Domains

JAK2 Recombinant Human Protein, JH1 and JH2 Domains

Brand:  Invitrogen PV4393

Additional Details : Weight : 0.00900kg

Product Code. 11515026

  • 11110.00 SEK / 10µg
Estimated Shipment: 14-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.


Description

Description

JAK2 is a protein tyrosine kinase involved in a specific subset of cytokine receptor signaling pathways and is mutated in polycythemia vera. This kinase contains both the JH1 (catalytic) and JH2 (pseudokinase) domains of JAK2.
Specifications

Specifications

NP_004963
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay
JAK2 Recombinant Human Protein
–80°C
1 Each
JAK2 Recombinant Human Protein
JAK
JAK2 Recombinant Human Protein
Human
Baculovirus
Cytoplasmic tyrosine kinase
0.32mg/mL
97.8kDa
10μg
For research use only. Not for use in diagnostic procedures
JAK2 JH1 JH2
JAK2 JH1 JH2 was pre-diluted in enzyme dilution buffer and assayed in 25mM HEPES (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL AXLtide peptide substrate (KKSRGDYMTMQIG) and trace [32P]-γ-ATP for 10 minutes at 30°C
Amino acids 532-1132
390nmoles of phosphate transferred to AXLtide peptide substrate (KKSRGDYMTMQIG) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2.08μg/mL
GST Tag
Kinase and inhibitors
90% by SDS-PAGE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.