Learn More
Invitrogen™ JAK2 [V617F] Recombinant Human Protein, JH1 and JH2 Domains
JAK2 is a protein tyrosine kinase involved in a specific subset of cytokine receptor signaling pathways.
Brand: Invitrogen™ PR8134A
Additional Details : Weight : 0.01000kg
Description
This kinase contains both the JH1 (catalytic) and JH2 (pseudokinase) domains of JAK2. The V617F mutation has been shown to be a cause of polycythemia vera.
Specifications
NP_004963 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
JAK2 JH1 JH2 V617F | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
97.9kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
JAK Kinase Family | |
Recombinant | |
Baculovirus |