Läs mer
Tocris Bioscience™ Janelia Fluor 525, NHS ester
Beskrivning
Application: Suitable for confocal microscopy and super resolution microscopy (SRM) techniques, including dSTORM (in both live and fixed cells). Janelia Fluor™ 525, NHS ester is cell permeable.
Properties and Photophysical Data: NHS ester can be converted to relevant substrate for use in self-labeling tag systems, e.g. HaloTag™ and SNAP-tag™. Can be multiplexed for two color imaging with Janelia Fluor™ 635, NHS ester (Cat. No. 6419). Excitation and emission maxima (λ) are 525 nm and 549 nm, respectively; quantum yield = 0.91; extinction coefficient = 122,000 M-1cm-1; A280 correction factor = 0.185; lactone-zwitterion equilibrium constant (KL-Z) = 0.068.
Please see the protocol for further information on protein/antibody labeling and conjugation.
We also offer Janelia Fluor™ conjugated antibodies and custom conjugation services .
HaloTag is a trademark of Promega Corporation, and SNAP-tag is a trademark of New England BioLabs, Inc.
Janelia Fluor is a registered trademark of Howard Hughes Medical Institute.
This product has been recently renamed. The previous name for this product was Janelia Fluor™ 525, SE.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 3,6-Di-1-(3,3-difluoroazetidinyl)-9-[2-carboxy-5-[[(2,5-dioxo-1-pyrrolidinyl)oxy]carbonyl]phenyl]xanthylium, inner salt |
| CAS | 2975167-21-4 |
| Kvantitet | 1 mg |
| Mål | Amine-Reactive Janelia Fluor Dyes |
| Molekylformel | C31H21F4N3O7 |
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.