Läs mer
Tocris Bioscience™ Janelia Fluor™ 630b, Maleimide
Beskrivning
Key Information: Janelia Fluor™ 630b, Maleimide is a spontaneously blinking fluorescent dye, supplied with a maleimide reactive group for conjugation (thiol reactivity).
Used for: suitable to characterize the physical properties of dense biomolecule condensates and complex macromolecular assemblies.
Application: suitable for super resolution microscopy (SRM) including single-molecule localization microscopy (SMLM).
Properties and Photophysical Data: Excitation and emission maxima (λ) are 651 nM and 666 nM, respectively.
Janelia Fluor is a registered trademark of Howard Hughes Medical Institute.
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-(2-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)-5,5-dimethyl-3,7-bis(3-(methylsulfonyl)azetidin-1-yl)-3'H,5H-spiro[dibenzo[b,e]siline-10,1'-isobenzofuran]-6'-carboxamide |
| Kvantitet | 200 μg |
| Mål | Sulfhydyl-Reactive Janelia Fluor Dyes |
| Molekylformel | C37H40N4O8S2Si |
| Renhet | 0.9 |